Learn More
Medchemexpress LLC HY-103483 5mg Medchemexpress, MD2-IN-1 CAS:111797-22-9 Purity:>98%
SDP
Catalog No. 501968122
Encompass_Preferred
Medchemexpress, HY-103483 5mg MD2-IN-1 CAS:111797-22-9 MD2-IN-1 is an inhibitor of Myeloid differentiation protein 2 (MD2) with a KD of 189  μM for the recombinant human MD2 (rhMD2). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8122 Supplier Medchemexpress LLC Supplier No. HY1034835MG
Medchemexpress, HY-103483 5mg MD2-IN-1 CAS:111797-22-9 MD2-IN-1 is an inhibitor of Myeloid differentiation protein 2 (MD2) with a KD of 189  μM for the recombinant human MD2 (rhMD2). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.