Learn More
Medchemexpress LLC MD2-TLR4-IN-1 | 2249801-12-3 | 98.2% | 10 MG
SDP
Catalog No. 5000340845
Encompass_Preferred
MD2-TLR4-IN-1 is a myeloid differentiation protein 2/toll-like receptor 4 (MD2-TLR4) complex antagonist. It inhibits Lipopolysaccharides (LPS)-induced expression of TNF-α and IL-6 in macrophages with IC50 values of 0.89 μM and 0.53 μM, respectively. This compound can be utilized for the study of acute lung injury (ALI).
  • Exerts anti-inflammatory activity by inhibiting NF-κB in RAW264.7 macrophages.
  • Directly binds to the MD2-TLR4 complex and disrupts MD2-TLR4 activation.
  • Suppresses NF-κB p65 subunit nuclear levels in LPS-induced macrophages.
  • Significantly inhibits macrophage infiltration and ameliorates histopathological changes in lung tissues.
  • Effectively reduces airspace inflammation and amends pulmonary lobule tissue structure.
  • Decreases LPS-induced myeloperoxidase (MPO) activity and serum TNF-α levels.
  • Reduces mRNA levels of TNF-α, IL-6, IL-1β, IL-12, IL-33, and COX-2 in lung tissues.
Catalog No. 50-003-40845 Supplier Medchemexpress LLC Supplier No. HY12859810MG
MD2-TLR4-IN-1 is a myeloid differentiation protein 2/toll-like receptor 4 (MD2-TLR4) complex antagonist. It inhibits Lipopolysaccharides (LPS)-induced expression of TNF-α and IL-6 in macrophages with IC50 values of 0.89 μM and 0.53 μM, respectively. This compound can be utilized for the study of acute lung injury (ALI).
  • Exerts anti-inflammatory activity by inhibiting NF-κB in RAW264.7 macrophages.
  • Directly binds to the MD2-TLR4 complex and disrupts MD2-TLR4 activation.
  • Suppresses NF-κB p65 subunit nuclear levels in LPS-induced macrophages.
  • Significantly inhibits macrophage infiltration and ameliorates histopathological changes in lung tissues.
  • Effectively reduces airspace inflammation and amends pulmonary lobule tissue structure.
  • Decreases LPS-induced myeloperoxidase (MPO) activity and serum TNF-α levels.
  • Reduces mRNA levels of TNF-α, IL-6, IL-1β, IL-12, IL-33, and COX-2 in lung tissues.