Learn More
Medchemexpress LLC HY-128598 5mg Medchemexpress, MD2-TLR4-IN-1 CAS:2249801-12-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1285985MG
Encompass_Preferred
Medchemexpress, HY-128598 5mg MD2-TLR4-IN-1 CAS:2249801-12-3 MD2-TLR4-IN-1 (compound 22m) is an inhibitor of myeloid differentiation protein 2/toll-like receptor 4 (MD2-TLR4) complex, inhibiting lipopolysaccharides (LPS)-induced expression of tumor necrosis factor alpha (TNF-α) and interleukin-6 (IL-6) in macrophages with IC50 values of 0.89 μM and 0.53 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9755