Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond MD50 Colorimeter: Chlorine HR

Catalog No. 01288087 Shop All Tintometer Group Products
Change view
Click to view available options
Reagent Form:
Tablet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Reagent Form
01-288-087 Tablet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-288-087 Supplier Lovibond Supplier No. 280140
Only null left
Add to Cart
Add to Cart

Portable colorimeter for measuring high levels of total chlorine in industrial water samples using the KI method.

  • Utilizes tablet chemistries for easy and exact reagent dosing
  • Rubberized side panels increase the durability of the instrument
  • Automatically stores the last 100 readings with time and date stamp
  • Advanced optical system reduces measurement time to less than 3 seconds per analysis
  • Robust Design: The MD50 is built to last. Its rugged industrial design, complete with rubber-protected sides and an IP67 rating, ensures unmatched durability and reliability even in demanding environments.
  • Accurate, Reproducible Results: The innovative optical system of the MD50 delivers exceptional accuracy and speed, ensuring precise readings in record time.
  • Factory Programmed Methods: All MD50 instruments come with pre-programmed methods for different types of reagents. Simply select the test you need, prepare your sample, and test! To simplify test selection, you can choose the reagent type from the settings and display only your desired methods in the methods list.
  • User-Defined Method: The MD50 also allows you to program a user-defined method using PC software.
  • One-Time-Zero Function: Save time by zeroing the instrument once for each sample. When testing a new sample, zero the instrument once, and all subsequent tests of that sample do not require re-zeroing. You can also activate or deactivate the OTZ function for all methods in the settings.
  • Automatic Data Storage: The MD50 automatically stores the last 100 readings and protocols with a time and date stamp. Results remain in storage even during a battery exchange.
  • Data Transfer: The MD50 offers smooth and effortless data transfer. You can transfer data to the AquaLX™ app via NFC or use a USB cable to transfer data to PC software. Data can also be exported in CSV format for programs like Excel.
  • Ergonomic Design: Whether you're using the MD50 in the field or in the lab, its design makes it easy to handle and view the display. In the field, the compact size and side protections ensure a firm grip during use. In the lab, rubber feet on the bottom of the housing eliminate the need for two-handed operation or chasing your instrument around the workbench.
  • GLP Compliance: To ensure data quality and integrity, the MD50 creates and saves a protocol for each change made during operation.
  • 2-year Warranty

Specifications

Backlight Option Yes
Battery Life Approx. 5000 Tests
Calibration Factory Calibrated
Certifications/Compliance CE
Connections USB-C, NFC
Cuvette Details Round, 24 mm, Glass
Data Storage 100 Data Records, Internal Ring Memory
Battery Type 2 x AA
Detector Type Photodiode Array
Display Graphic LCD
IP Rating IP67
Languages English
Light Source LED
Spectral Bandwidth Not Measured
Type Chlorine Colorimeter
Voltage 1.5 V
Wavelength Selection Automatic
For Use With (Application) Water Testing, Non-labortatory Setting
Measurement Range 5 to 200 mg/L Cl2, 0.2 to 16 % NaOCl
Model MD50 Colorimeter, Chlorine HR (KI)
Photometric Accuracy 3% FS (20°C to 25°C)
Photometric Range 0 to 3 Abs
Quantity 1 Ea.
Reagent Form Tablet
Sample Size 10 mL
Test Method Colorimetric, Potassium Iodide (KI)
Warranty 2 Years for Manufacturing Defects
Wavelength 530 nm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.