Learn More
Inmark Inc MD8200V12 - EXAKT-PAK UNI-PAK
SDP
Catalog No. NC3441609
Encompass_Preferred
  • Insulated shipping solution designed for biological substances
  • Maintains cool temperature for up to 24 hours
  • Complies with USDOT Regulations and IATA Packing Instruction 650
  • Features corrugated outer box with 1 in insulation
  • Includes leakproof bags and foam inserts
  • Preprinted with necessary labels for Biological Substance, Category B, and UN3373
  • Suitable for transporting blood tubes, urine tubes, and vials
  • Includes absorbent material for safety
  • Recommended for specimen transport
  • Compatible with various receptacles such as tubes, jars, and vials
Catalog No. NC3441609 Supplier Inmark Inc Supplier No. 26994
  • Insulated shipping solution designed for biological substances
  • Maintains cool temperature for up to 24 hours
  • Complies with USDOT Regulations and IATA Packing Instruction 650
  • Features corrugated outer box with 1 in insulation
  • Includes leakproof bags and foam inserts
  • Preprinted with necessary labels for Biological Substance, Category B, and UN3373
  • Suitable for transporting blood tubes, urine tubes, and vials
  • Includes absorbent material for safety
  • Recommended for specimen transport
  • Compatible with various receptacles such as tubes, jars, and vials