Learn More
Medchemexpress LLC HY-15451 5mg Medchemexpress, MDA 19 CAS:1048973-47-2 Purity:>98%
SDP
Catalog No. 502081726
Encompass_Preferred
Medchemexpress, HY-15451 5mg MDA 19 CAS:1048973-47-2 MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. MDA 19 has antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1726 Supplier Medchemexpress LLC Supplier No. HY154515MG
Medchemexpress, HY-15451 5mg MDA 19 CAS:1048973-47-2 MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. MDA 19 has antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.