Learn More
Medchemexpress LLC MDA 19 (BZO-HEXOXIZID) | 1048973-47-2 | C21H23N3O2 | 50 MG
SDP
Catalog No. 5000376803
Encompass_Preferred
MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. It exhibits antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity.
  • Potent and selective agonist of human cannabinoid receptor 2 (CB2)
  • Exhibits antiallodynic effects in a rat model of neuropathic pain
  • Does not affect rat locomotor activity
  • Higher affinity at human CB2 (Ki = 43.3 nM) than human CB1
  • Nearly 70-fold higher affinity at rat CB2 (Ki = 16.3 nM) than rat CB1
Catalog No. 50-003-76803 Supplier Medchemexpress LLC Supplier No. HY1545150MG
MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. It exhibits antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity.
  • Potent and selective agonist of human cannabinoid receptor 2 (CB2)
  • Exhibits antiallodynic effects in a rat model of neuropathic pain
  • Does not affect rat locomotor activity
  • Higher affinity at human CB2 (Ki = 43.3 nM) than human CB1
  • Nearly 70-fold higher affinity at rat CB2 (Ki = 16.3 nM) than rat CB1