Learn More
Cayman Chemical MDA 19, Purity: ≥95%, CAS: 1048973-47-2, 10 mg
SDP
Catalog No. 502887280
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid; has been found in illicit substances seized by law enforcement; reduces tactile allodynia in rats and mice; intended for research and forensic applications
Catalog No. 50-288-7280 Supplier Cayman Chemical Supplier No. 1056310 MG
An analytical reference standard categorized as a synthetic cannabinoid; has been found in illicit substances seized by law enforcement; reduces tactile allodynia in rats and mice; intended for research and forensic applications