Learn More
Enzo Life Sciences ALDetect (MDA-specific) Lipid Peroxidation assay kit (1x25tests)
SDP
Catalog No. NC0804247
Encompass_Preferred
Applications: Colorimetric detection. Contents: Reagent R1 (Prod. No. BML-KI527) (3x18ml;N-methyl-2-phenylindole, in acetonitrile). Reagent R2 (Prod. No. BML-KI606) (1x16.5ml; Conc. Hydrochloric acid). MDA Standard (Prod. No. BML-KI529) (1ml; 1,1,3,3-tetramethoxypropane (TMOP) in Tris-hydrochloric acid). BHT (Prod. No. BML-KI530) (2ml; Butylated hydroxytoluene in acetonitrile). Probucol (Prod. No. BML-KI531) (1.1ml; Probucol in methanol). Methanol (Prod. No. BML-KI532) (30ml)
Catalog No. NC0804247 Supplier Enzo Life Sciences Supplier No. BMLAK1710001
Add to Cart
Add to Cart
Applications: Colorimetric detection. Contents: Reagent R1 (Prod. No. BML-KI527) (3x18ml;N-methyl-2-phenylindole, in acetonitrile). Reagent R2 (Prod. No. BML-KI606) (1x16.5ml; Conc. Hydrochloric acid). MDA Standard (Prod. No. BML-KI529) (1ml; 1,1,3,3-tetramethoxypropane (TMOP) in Tris-hydrochloric acid). BHT (Prod. No. BML-KI530) (2ml; Butylated hydroxytoluene in acetonitrile). Probucol (Prod. No. BML-KI531) (1.1ml; Probucol in methanol). Methanol (Prod. No. BML-KI532) (30ml)