Learn More
Cayman Chemical MDA-19 4-hydroxybenzoyl metabolite, Purity: ≥98%, 5 mg
SDP
Catalog No. 502911633
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential metabolite of MDA-19; intended for research and forensic applications
Catalog No. 50-291-1633 Supplier Cayman Chemical Supplier No. 360405 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential metabolite of MDA-19; intended for research and forensic applications