Learn More
AdipoGen anti-MDA5 (human), mAb (Hely-1) (100 µg)
SDP
Supplier:  AdipoGen AG20B0013C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human MDA5. Isotype: Mouse IgG1. Clone: Hely-1. Applications: ELISA, IP, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. RIG-I and MDA5 are highly conserved helicases involved in the innate immune response to virus. RIG-I is a member of the DEAD-box RNA helicases and is activated by cytoplasmic dsRNA and 5’-ppp RNA produced during the viral replication. The protein is characterized by a N-terminal region with two caspase recruitment domains (CARD) and a C-terminal region harboring potential ATP-dependent RNA helicase activity. RIG-1 recruits the CARD adaptor inducing IFN-beta (Cardif) in a CARD-CARD-dependent manner resulting in NF-kappaB and IRF3 activation.
Catalog No. 50-205-2910