Learn More
Enzo Life Sciences MDA5 (human) polyclonal antibody (AT113) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX210935C100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human MDA5 (melanoma differentiation-associated gene 5) (aa 78-555). UniProt: Q9BYX4. Source: Purified from rabbit serum. Species Reactivity: Human. Applications: IP, WB. Recommended dilutions: Immunoprecipitation (1:100). Western Blot (1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. NC0907169