Learn More
Enzo Life Sciences MDA5 monoclonal antibody (1B6) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX804878C100
Encompass_Preferred
Clone: 1B6. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human MDA5 (melanoma differentiation-associated gene 5) (aa 78-555). UniProt: Q9BYX4. Source: Purified from hybridoma tissue culture supernatant. Species Reactivity: Human, Mouse. Applications: ELISA, WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. NC0907132