Learn More
Enzo Life Sciences MDA5 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2990100
Encompass_Preferred
Isotype: IgG. Immunogen: Synthetic peptide corresponding to 16 amino acids from near the center of human MDA5. Species reactivity: Human, Mouse. Applications: ELISA, IF, IHC (PS), WB. Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-2838