Learn More
Teknova MDAG-1 Media. 500mL, Sterile. 8 pack.
SDP
Catalog No. 502387683
Encompass
25mM Disodium Phosphate,25mM Monopotassium Phosphate,50mM Ammonium Chloride,5mM Sodium Sulfate,2mM Magnesium Sulfate,0.2X Trace Metals,0.1% Cerelose Anhydrous Dextrose (Glucose),0.1% Aspartate,200g/mL Amino Acids, Minus Cysteine and Tyrosine
Catalog No. 50-238-7683 Supplier Teknova Supplier No. M41078PK
25mM Disodium Phosphate,25mM Monopotassium Phosphate,50mM Ammonium Chloride,5mM Sodium Sulfate,2mM Magnesium Sulfate,0.2X Trace Metals,0.1% Cerelose Anhydrous Dextrose (Glucose),0.1% Aspartate,200g/mL Amino Acids, Minus Cysteine and Tyrosine