Learn More
ATCC MDCK.2; Kidney; Dog (Canis familiaris)
Catalog No. 502382770
Encompass
MDCK.2 is an epithelial-like cell that was isolated from the distal renal tubule of a dog. This cell line was deposited by Y. Reid, E. Cedrone, E-Eckard-Amar, and can be used in toxicology research.
Catalog No. 50-238-2770 Supplier ATCC Supplier No. CRL2936
Add to Cart
Add to Cart
MDCK.2 is an epithelial-like cell that was isolated from the distal renal tubule of a dog. This cell line was deposited by Y. Reid, E. Cedrone, E-Eckard-Amar, and can be used in toxicology research.