Learn More
Medchemexpress LLC MDEG-541 | 98.1% | 674.83 g·mol⁻¹ | C35H38N4O6S2 | 10 MG
SDP
Catalog No. 5000287242
Encompass_Preferred
MDEG-541 is a PROTAC small-molecule degrader that targets the MYC-MAX dimerization complex and exhibits antiproliferative activity by reducing cellular levels of MYC, GSPT1, GSPT2, and PLK1. It is provided as a research reagent with reported physicochemical characterization and recommended storage conditions.
  • Potent MYC-MAX degrader with PROTAC mechanism of action.
  • Demonstrates antiproliferative activity and downregulates MYC, GSPT1, GSPT2, and PLK1.
  • High reported purity suitable for research applications (≈98.1%).
  • Supplied in small research quantities for biochemical and cellular studies.
  • Stable as powder when stored at -20°C; follow solvent storage guidelines for dissolved material.
Catalog No. 50-002-87242 Supplier Medchemexpress LLC Supplier No. HY15025910MG
MDEG-541 is a PROTAC small-molecule degrader that targets the MYC-MAX dimerization complex and exhibits antiproliferative activity by reducing cellular levels of MYC, GSPT1, GSPT2, and PLK1. It is provided as a research reagent with reported physicochemical characterization and recommended storage conditions.
  • Potent MYC-MAX degrader with PROTAC mechanism of action.
  • Demonstrates antiproliferative activity and downregulates MYC, GSPT1, GSPT2, and PLK1.
  • High reported purity suitable for research applications (≈98.1%).
  • Supplied in small research quantities for biochemical and cellular studies.
  • Stable as powder when stored at -20°C; follow solvent storage guidelines for dissolved material.