Learn More
Medchemexpress LLC MDEG-541 | C₃₅H₃₈N₄O₆S₂ | 50 MG
SDP
Catalog No. 5000375579
Encompass_Preferred
MDEG-541 is a potent MYC-MAX degrader. It is a PROTAC based on the MYC-MAX dimerization inhibitor 10058-F4 derivative 28RH and Thalidomide. It demonstrates antiproliferative activity and reduces the expression of GSPT1, MYC, GSPT2, and PLK1 proteins.
  • Decreases the expression of GSPT1, MYC, GSPT2, and PLK1 proteins in a time-dependent manner in KP4 cells.
  • Inhibits cell viability in HCT116 and PSN1 cells.
  • Reduces MYC expression in a dose-dependent manner in HCT116 and PSN1 cells.
Catalog No. 50-003-75579 Supplier Medchemexpress LLC Supplier No. HY15025950MG
MDEG-541 is a potent MYC-MAX degrader. It is a PROTAC based on the MYC-MAX dimerization inhibitor 10058-F4 derivative 28RH and Thalidomide. It demonstrates antiproliferative activity and reduces the expression of GSPT1, MYC, GSPT2, and PLK1 proteins.
  • Decreases the expression of GSPT1, MYC, GSPT2, and PLK1 proteins in a time-dependent manner in KP4 cells.
  • Inhibits cell viability in HCT116 and PSN1 cells.
  • Reduces MYC expression in a dose-dependent manner in HCT116 and PSN1 cells.