Learn More
Medchemexpress LLC MDEG-541 | C35H38N4O6S2 | 25 MG
SDP
Catalog No. 5000374383
Encompass_Preferred
MDEG-541 is a potent MYC-MAX degrader that functions as a PROTAC (Proteolysis-targeting chimera). It is derived from a MYC-MAX dimerization inhibitor and Thalidomide. This compound exhibits antiproliferative activity and reduces the expression of specific proteins.
  • Potent MYC-MAX degrader
  • Functions as a PROTAC
  • Exhibits antiproliferative activity
  • Reduces the expression of GSPT1, MYC, GSPT2, and PLK1 proteins
  • Soluble in DMSO
  • Appears as a solid with a light yellow to yellow color
  • Stable for 3 years as powder at -20°C
  • Stable for 6 months in solvent at -80°C
  • Stable for 1 month in solvent at -20°C
  • Inhibits cell viability in HCT116 and PSN1 cells
Catalog No. 50-003-74383 Supplier Medchemexpress LLC Supplier No. 25MGHY150259
Add to Cart
Add to Cart
MDEG-541 is a potent MYC-MAX degrader that functions as a PROTAC (Proteolysis-targeting chimera). It is derived from a MYC-MAX dimerization inhibitor and Thalidomide. This compound exhibits antiproliferative activity and reduces the expression of specific proteins.
  • Potent MYC-MAX degrader
  • Functions as a PROTAC
  • Exhibits antiproliferative activity
  • Reduces the expression of GSPT1, MYC, GSPT2, and PLK1 proteins
  • Soluble in DMSO
  • Appears as a solid with a light yellow to yellow color
  • Stable for 3 years as powder at -20°C
  • Stable for 6 months in solvent at -80°C
  • Stable for 1 month in solvent at -20°C
  • Inhibits cell viability in HCT116 and PSN1 cells