Learn More
Medchemexpress LLC Mdeg-541 | C35H38N4O6S2 | 5 MG
SDP
Catalog No. 5000375316
Encompass_Preferred
MDEG-541 is a potent MYC-MAX degrader and a PROTAC, derived from the MYC-MAX dimerization inhibitor 10058-F4 (derivative 28RH) and Thalidomide (HY-14658). It exhibits antiproliferative activity and reduces the expression of GSPT1, MYC, GSPT2, and PLK1 proteins. It also decreases the expression of MYC in a dose-dependent manner and inhibits cell viability in HCT116 and PSN1 cells.
  • Potent MYC-MAX degrader
  • PROTAC based on 10058-F4 derivative and Thalidomide
  • Shows antiproliferative activity
  • Decreases expression of GSPT1, MYC, GSPT2, PLK1 protein
  • Inhibits cell viability in HCT116 and PSN1 cells
Catalog No. 50-003-75316 Supplier Medchemexpress LLC Supplier No. HY1502595MG
MDEG-541 is a potent MYC-MAX degrader and a PROTAC, derived from the MYC-MAX dimerization inhibitor 10058-F4 (derivative 28RH) and Thalidomide (HY-14658). It exhibits antiproliferative activity and reduces the expression of GSPT1, MYC, GSPT2, and PLK1 proteins. It also decreases the expression of MYC in a dose-dependent manner and inhibits cell viability in HCT116 and PSN1 cells.
  • Potent MYC-MAX degrader
  • PROTAC based on 10058-F4 derivative and Thalidomide
  • Shows antiproliferative activity
  • Decreases expression of GSPT1, MYC, GSPT2, PLK1 protein
  • Inhibits cell viability in HCT116 and PSN1 cells