Learn More
3B Scientific™ MDF™ Taylor Hammer, Black
SDPSCI_ED
Catalog No. S20682 Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
S20682 1
1 options
Catalog No. S20682 Supplier 3B Scientific™ Supplier No. W78011
Add to Cart
Add to Cart
3B Scientific™ MDF™ Taylor Hammer incorporates modern design and functionality for eliciting myotatic and plantar responses with less effort and greater patient comfort.
  • Silicone triangular head has a beveled apex and base to elicit myotatic reflexes
  • Soft base and rounded apex are perfectly suited for provoking myotatic responses
  • Suited to obtain chest sounds and to percuss the abdomen
  • Weighted chrome-plated zinc-alloy handle is precisely balanced for increased control of percussion force
  • Handle is also designed with a pointed tip for eliciting cutaneous and plantar reflexes.
Specifications
Product Type Therapy Equipment
Includes Hammer
Width (Metric) 5.6 cm
Height (Metric) 2.5 cm
Quantity 1
Material Plastic, Metal
Color Black
Languages English