Learn More
Electron Microscopy Sciences Modified Mayer's Hematoxylin | 500ml
SDP
Catalog No. 501928514
Encompass_Preferred
Modified Mayer's Hematoxylin stock solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It contains Hematoxylin 0.4%, Ammonium Aluminum Sulfate 0.5%, Sodium Iodate 0.03%, Citric Acid 0.1%, and Chloral Hydrate 5%.
Catalog No. 50-192-8514 Supplier Electron Microscopy Sciences Supplier No. 2604318
Modified Mayer's Hematoxylin stock solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It contains Hematoxylin 0.4%, Ammonium Aluminum Sulfate 0.5%, Sodium Iodate 0.03%, Citric Acid 0.1%, and Chloral Hydrate 5%.