Learn More
Aviva Systems Biology Corp MDFIC Antibody - middle region (ARP50878_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50878_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MDFIC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MDFIC. Alternate names: HICApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-60461