Learn More
Aviva Systems Biology Corp MDFIC Antibody - N-terminal region (ARP50877_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50877_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MDFIC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MDFIC. Alternate names: HICApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-014-60460