Learn More
Aviva Systems Biology Corp MDH1 Antibody - middle region (ARP48284_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48284_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MDH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MDH1. Alternate names: MDH-s, MDHA, MGC:1375, MOR2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59784