Learn More
ECM Bioscience mDia2 (C-terminal region) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience DP3491
Encompass_Preferred
This antibody was affinity purified using mDia2 (C-terminal region) peptide (without carrier). The antibody detects a 135 kDa* protein corresponding to the apparent molecular mass of mDia2 on SDS-PAGE immunoblots of various cells and tissues, including human A431 and HeLa cells, mouse brain tissue and C2C12 cells, rat PC12 cells, and rabbit spleen fibroblasts. The immunogen was mDia2 synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal region of mouse mDia2 (Diap3). This peptide sequence is highly conserved in rat and human mDia2, and has low homology to other formins. Accession #Q9Z207. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Diap3, Dia2, Drf3, formin
Catalog No. 50-172-0755