Learn More
MEDILUMINE INC MdiLumine Inc. AkaLumine HCL (TokeOni), 5 mg
Catalog No. NC3968105
Encompass_Preferred
Luciferin analog displaying luminescence peak at 677 nm (near-infrared window) with standard firefly luciferase reporter system enabling deeper and more sensitive tissue imaging due to the reduced absorption and scattering of light compared to photonic emissions in the visible wavelengths. The analog also provides stroger emissions for for in vitro assays compared to use of D-Luciferin.
Catalog No. NC3968105 Supplier MEDILUMINE INC Supplier No. 305AKA
Luciferin analog displaying luminescence peak at 677 nm (near-infrared window) with standard firefly luciferase reporter system enabling deeper and more sensitive tissue imaging due to the reduced absorption and scattering of light compared to photonic emissions in the visible wavelengths. The analog also provides stroger emissions for for in vitro assays compared to use of D-Luciferin.