Learn More
Apexbio Technology LLC Mdivi 1, 338967-87-6, MFCD00974506, 10 mg
SDP
Supplier:  Apexbio Technology LLC A447210
Encompass_Preferred
MF: C15H10Cl2N2O2S, Solubility: 17.65mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. Mdivi-1 is a selective cell-permeable inhibitor of mitochondrial division DRP1 (dynamin-related GTPase) and mitochondrial division Dynamin I (Dnm1). DRP1, a member of the dynamin family of large GTPases, mediates mitochondrial fission.
Catalog No. 50-101-2022