Learn More
Medchemexpress LLC HY-15886 10mg , Mdivi-1 CAS:338967-87-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1588610MG
Encompass_Preferred
HY-15886 10mg Mdivi-1 CAS: 338967-87-6 Mdivi-1 is a selective dynamin-related protein 1 (Drp1) inhibitor. Mdivi-1 is a mitochondrial division/mitophagy inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9150