Learn More
Selleck Chemical LLC Mdivi-1 50mg 338967-87-6
SDP
Supplier:  Selleck Chemical LLC S716250MG
Encompass_Preferred
Mdivi-1 is a selective cell-permeable inhibitor of mitochondrial division DRP1 (dynamin-related GTPase) and mitochondrial division Dynamin I (Dnm1) with IC50 of 1-10 ?M. Mdivi-1 attenuates mitophagy and enhances apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4471