Learn More
Aviva Systems Biology Corp MDK Antibody - C-terminal region (ARP63426_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63426_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MDK. It was validated on Western Blot. . Alternate names: FLJ27379, MK, NEGF2ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-965