Learn More
American Radiolabeled Chemicals Inc MDL 100907 ME-3H 50UC
SDP
Catalog No. 50352955
Encompass_Preferred
MDL 100907-methyl-3H 50uCi in ethanol, 1 mCi/ml, 70-87 Ci/mmol, M.W. 373.47, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-955 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 194150UC
Add to Cart
Add to Cart
Request a Quote
MDL 100907-methyl-3H 50uCi in ethanol, 1 mCi/ml, 70-87 Ci/mmol, M.W. 373.47, Shipped in Dry Ice, Exclusive