Learn More
Cayman Chemical MDL 12330AhydrochlorIde 10mg
SDP
Catalog No. 502969375
Encompass_Preferred
An adenylyl cyclase inhibitor that can also inhibit cAMP and cGMP phosphodiesterases and block slow extracellular and store-operated Ca2+ entry into cells; blocks voltage-dependent K+ channels, extends action potential duration, and enhances insulin secretion in pancreatic β-cells
Catalog No. 50-296-9375 Supplier Cayman Chemical Supplier No. 1455910 MG
An adenylyl cyclase inhibitor that can also inhibit cAMP and cGMP phosphodiesterases and block slow extracellular and store-operated Ca2+ entry into cells; blocks voltage-dependent K+ channels, extends action potential duration, and enhances insulin secretion in pancreatic β-cells