Learn More
TARGETMOL CHEMICALS INC MDL-28170 25MG
SDP
Catalog No. 502577114
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MDL-28170 (Calpain Inhibitor III) is a Cysteine protease. purity: 98%
Catalog No. 50-257-7114 Supplier TARGETMOL CHEMICALS INC Supplier No. T247025MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MDL-28170 (Calpain Inhibitor III) is a Cysteine protease. purity: 98%