Learn More
Enzo Life Sciences MDL 28170 (50mg). CAS: 88191-84-8
SDP
Catalog No. 502011470
Encompass_Preferred
A potent and selective cell permeable calpain inhibitor. Inhibits oxidative damage-induced apoptosis in PC12 cells and capsaicin-induced apoptosis in dorsal root ganglion neurons. It protects against anoxia in rat optic nerves and reduces neuronal damage in an animal model of global cerebral ischemia(50 mg/kg). In isolated hippocampal pyramidal neurons, it inhibits Ca2+-induced suppression of neurite outgrowth (complete inhibition at 20 µM). Calpain I Ki = 8 nM; Cathepsin B Ki = 24 nM in isolated enzyme assays. IC50 = 0.3 µM in intact cell assay. Alternative name: N-Benzyloxycarbonylvalylphenylalaninal. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (200mM). Long Term Storage: -20°C.
Catalog No. 50-201-1470 Supplier Enzo Life Sciences Supplier No. BMLPI1300050
A potent and selective cell permeable calpain inhibitor. Inhibits oxidative damage-induced apoptosis in PC12 cells and capsaicin-induced apoptosis in dorsal root ganglion neurons. It protects against anoxia in rat optic nerves and reduces neuronal damage in an animal model of global cerebral ischemia(50 mg/kg). In isolated hippocampal pyramidal neurons, it inhibits Ca2+-induced suppression of neurite outgrowth (complete inhibition at 20 µM). Calpain I Ki = 8 nM; Cathepsin B Ki = 24 nM in isolated enzyme assays. IC50 = 0.3 µM in intact cell assay. Alternative name: N-Benzyloxycarbonylvalylphenylalaninal. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (200mM). Long Term Storage: -20°C.