Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MDL-28170 (Calpain inhibitor III) | 88191-84-8 | MFCD00798796 | >90.0% | 10 MG
SDP

Catalog No. 5000298791
Encompass_Preferred

MDL-28170 is a potent, selective, membrane-permeable cysteine protease inhibitor of calpain that rapidly penetrates the blood-brain barrier and also inhibits γ-secretase. It is commonly used in neurological and inflammation research to probe protease-dependent signaling and neurodegenerative processes.

  • Potent, selective calpain inhibitor.
  • Membrane-permeable and blood-brain barrier penetrant.
  • Dual activity: calpain inhibition and γ-secretase blockade.
  • Applicable to neurological disease and inflammation research.
  • Supplied as a powder; store at -20°C for long-term stability.
  • Molecular formula C22H26N2O4; molecular weight 382.45 g/mol.
  • CAS number 88191-84-8.

Catalog No. 50-002-98791 Supplier Medchemexpress LLC Supplier No. HY1823610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MDL-28170 is a potent, selective, membrane-permeable cysteine protease inhibitor of calpain that rapidly penetrates the blood-brain barrier and also inhibits γ-secretase. It is commonly used in neurological and inflammation research to probe protease-dependent signaling and neurodegenerative processes.

  • Potent, selective calpain inhibitor.
  • Membrane-permeable and blood-brain barrier penetrant.
  • Dual activity: calpain inhibition and γ-secretase blockade.
  • Applicable to neurological disease and inflammation research.
  • Supplied as a powder; store at -20°C for long-term stability.
  • Molecular formula C22H26N2O4; molecular weight 382.45 g/mol.
  • CAS number 88191-84-8.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.