Learn More
Electron Microscopy Sciences ThermoSafe Model 353 EPS Foam Mailer for Cans
SDP
Catalog No. 5034565
Encompass_Preferred
Accommodates the  most widely used laboratory collection tubes, vials, and bottles. Helps to prevent breakage during handling and shipment. Seamless construction with lined screw-cap to prevent leakage. completely autoclavable. EPS foam mailers and mailing sleeves available.
Diameter: 2"
OAL: 5.5"
O.D.: 6-1/4 x 3 1/4 x 3-1/16"
Catalog No. 50-345-65 Supplier Electron Microscopy Sciences Supplier No. 6151853
Accommodates the  most widely used laboratory collection tubes, vials, and bottles. Helps to prevent breakage during handling and shipment. Seamless construction with lined screw-cap to prevent leakage. completely autoclavable. EPS foam mailers and mailing sleeves available.
Diameter: 2"
OAL: 5.5"
O.D.: 6-1/4 x 3 1/4 x 3-1/16"