Learn More
Enzo Life Sciences MDL-72222 (50mg). CAS: 40796-97-2
SDP
Supplier:  Enzo Life Sciences BMLNS5300050
Encompass_Preferred
Serotonin 5-HT3 antagonist. Alternative name: Bemesetron. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-1216