Learn More
Cayman Chemical MDL 72527 (hydrochloride), Purity: ≥95%, CAS: 93565-01-6, 10 mg
SDP
Catalog No. 502954321
Encompass_Preferred
A PAO inhibitor that inhibits spermine oxidase and N1-acetylpolyamine oxidase (IC50s = 6.1 and 0.02 µM, respectively); inhibits PAO-dependent H2O2 production in vitro and in vivo
Catalog No. 50-295-4321 Supplier Cayman Chemical Supplier No. 1399610 MG
A PAO inhibitor that inhibits spermine oxidase and N1-acetylpolyamine oxidase (IC50s = 6.1 and 0.02 µM, respectively); inhibits PAO-dependent H2O2 production in vitro and in vivo