Learn More
David Kopf Instruments David Kopf Instruments Model 940 Small Animal Stereotaxic, Digital Display Console, 80 mm Travel
SDP
Catalog No. NC2199087
Encompass
The David Kopf Instruments Model 940 Small Animal Stereotaxic Instrument with Digital Display Console is a precision tool designed for accurate placement in small animal research. It features robust construction, a digital display, and multi-dimensional adjustments with a 10 micron resolution. Its versatility, adaptability for various small animals, and capability for a full range of motion make it a standard choice for researchers, enabling enhanced accuracy and ease of use in experimental setups.
This item is not returnable. View return policy
Catalog No. NC2199087 Supplier David Kopf Instruments Supplier No. 940
Add to Cart
Add to Cart
Request a Quote
The David Kopf Instruments Model 940 Small Animal Stereotaxic Instrument with Digital Display Console is a precision tool designed for accurate placement in small animal research. It features robust construction, a digital display, and multi-dimensional adjustments with a 10 micron resolution. Its versatility, adaptability for various small animals, and capability for a full range of motion make it a standard choice for researchers, enabling enhanced accuracy and ease of use in experimental setups.