Learn More
Cell Systems Corporation Medium Kit Without Serum and With CultureBoost-R (4Z3-500-R)
SDP
Catalog No. NC1417959
Encompass_Preferred
Our base medium without serum can be customized to meet your specific cell culture needs. For complete medium, add 5-20% serum (defined FBS or equivalent) (not included) and the included CultureBoost-R supplement. This kit consists of 500mL Medium Without Serum, 10mL CultureBoost-R (containing human recombinant growth factors) and 10mL Attachment Factor . 4Z3-500-R is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.
Catalog No. NC1417959 Supplier Cell Systems Corporation Supplier No. 4Z3500R
Our base medium without serum can be customized to meet your specific cell culture needs. For complete medium, add 5-20% serum (defined FBS or equivalent) (not included) and the included CultureBoost-R supplement. This kit consists of 500mL Medium Without Serum, 10mL CultureBoost-R (containing human recombinant growth factors) and 10mL Attachment Factor . 4Z3-500-R is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.