Learn More
Abnova™ MDM2 CISH Probe
Catalog No. 89335567 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
Catalog No. For Use With (Application)
89-335-567 CISH-P
1 options
Catalog No. 89-335-567 Supplier Abnova™ Supplier No. CG0020
Add to Cart
Add to Cart
MDM2 CISH Probe is designed for the qualitative detection of human MDM2 gene in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
The probe is intended to be used in combination with the CISH Implementation Kit 1, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
Hybridization signals of digoxigenin-labeled polynucleotides appear as brown- to dark brown colored distinct dots (MDM2 gene region).
Normal situation: In interphases of normal cells or cells without an amplification involving the MDM2 gene locus, two distinct dot-shaped brown signals per nucleus appear.
Aberrant situation: In a cell with amplification of the MDM2 gene locus or polysomy of chromosome 12, multiple copies of the brown signal or brown signal clusters will be observed. Other signal distribution may be observed in some abnormal samples which might result in a different signal pattern than described above, indicating variant rearrangements.
Unexpected signal patterns should be further investigated.
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use.
For Use With (Application) CISH-P
Includes Reagent Provided:
1. Digoxigenin-labeled polynucleotides targeting sequences mapping in 12q15* (chr12:69,190,708-69,430,185) harboring the MDM2 gene
2. Formamide based hybridization buffer
*according to Human Genome Assembly GRCh37/hg19