Learn More
Medchemexpress LLC HY-130684 5mg Medchemexpress, MDM2-IN-1 CAS:1410737-09-5 Purity:>98%
SDP
Catalog No. 502081247
Encompass_Preferred
Medchemexpress, HY-130684 5mg MDM2-IN-1 CAS:1410737-09-5 MDM2-IN-1 (Compound 30) is a synthetic MDM2-p53 interaction (MDM2) inhibitor and contains the trans (D-)configuration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1247 Supplier Medchemexpress LLC Supplier No. HY1306845MG
Medchemexpress, HY-130684 5mg MDM2-IN-1 CAS:1410737-09-5 MDM2-IN-1 (Compound 30) is a synthetic MDM2-p53 interaction (MDM2) inhibitor and contains the trans (D-)configuration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.