Learn More
Aviva Systems Biology Corp MDM4 Antibody - N-terminal region (AVARP06007_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP06007_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MDM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MDM4. Alternate names: DKFZp781B1423, MDMX, MGC132766, MRP1, HDMXApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-62538