Learn More
Cayman Chemical MDMB-CHMICA metabolIte M2 1mg
SDP
Catalog No. 502984718
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of MDMB-CHMICA; intended for research and forensic applications
Catalog No. 50-298-4718 Supplier Cayman Chemical Supplier No. 193161 MG
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of MDMB-CHMICA; intended for research and forensic applications