Learn More
Cayman Chemical MDMB-4en-PICA butanoic acid metabolite, Purity: ≥98%, 5 mg
SDP
Catalog No. 502938126
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential metabolite of MDMB-4en-PICA based on the published metabolism of MDMB-PICA; intended for research and forensic applications
Catalog No. 50-293-8126 Supplier Cayman Chemical Supplier No. 289435 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential metabolite of MDMB-4en-PICA based on the published metabolism of MDMB-PICA; intended for research and forensic applications