Learn More
ATCC MDOK; Kidney; Sheep (Ovis aries)
Catalog No. 502381998
Encompass
MDOK is an epithelial-like cell that was isolated from the kidney of a male sheep. This cell line was deposited by RB Owens.
Catalog No. 50-238-1998 Supplier ATCC Supplier No. CRL1633
MDOK is an epithelial-like cell that was isolated from the kidney of a male sheep. This cell line was deposited by RB Owens.