Learn More
Medchemexpress LLC HY-14503 10mg Medchemexpress, MDR-1339 CAS:1018946-38-7 Purity:>98%
SDP
Catalog No. 502081606
Encompass_Preferred
Medchemexpress, HY-14503 10mg MDR-1339 CAS:1018946-38-7 MDR-1339 (DWK-1339) is an orally active and blood-brain-barrier-permeable Aβ-aggregation inhibitor, used in the research of Alzheimer's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1606 Supplier Medchemexpress LLC Supplier No. HY1450310MG
Medchemexpress, HY-14503 10mg MDR-1339 CAS:1018946-38-7 MDR-1339 (DWK-1339) is an orally active and blood-brain-barrier-permeable Aβ-aggregation inhibitor, used in the research of Alzheimer's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.