Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3 Percent Platinum on Activated Carbon Powder, Reduced, Water Wet, 22 microns, BASF Catalyst, Spectrum™ Chemical
SDP

Numéro de catalogue. 18607959 missing translation for 'brandsLinkText'
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
18-607-959 100 g
18-607-957 5 g
18-607-958 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 18-607-959 Fournisseur Spectrum Chemical Manufacturing Corporation Code fournisseur SP203100G
Get Up to $500 in Chemicals with Purchase
Afficher toutes les promotions
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
IUPAC Name platinum
SMILES [Pt]

Spécifications

Physical Form Powder or Liquid
Quantity 100 g
Grade Not Available
Packaging Amber Glass Bottle
Chemical Name or Material 3% Platinum on Activated Carbon Powder, Reduced, Water Wet, 22 μ, BASF Catalyst

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.