Learn More
Aviva Systems Biology Corp MDS032 Antibody - N-terminal region (ARP39361_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39361_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MDS032. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MDS032. Alternate names: P31, SLT1, MDS032ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-53137